ADVH49B829BD AChemBlock

benzyl 4-{[(benzyloxy)carbonyl]amino}-2,7-diazaspiro[4.4]nonane-2-carboxylate

  • Molecular Weight : 409.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-4-benzyloxy-carbonyl-amino-2-7-diazaspiro-4-4-nonane-2-carboxylate-308441.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85131
Availability details for
Quantity
  • Availability
  • Add to Cart