ADVH49B829BF AChemBlock

benzyl 2-[[(2S)-2-[[(2R)-2-[[2-(tert-butoxycarbonylamino)acetyl]amino]-3-phenyl-propanoyl]amino]-4-methyl-pentanoyl]amino]acetate

  • Molecular Weight : 582.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2-2s-2-2r-2-2-tert-butoxycarbonylamino-acetyl-amino-3-phenyl-propanoyl-amino-4-methyl-pentanoyl-amino-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85134
Availability details for
Quantity
  • Availability
  • Add to Cart