ADVH49B829C0 AChemBlock

benzyl 1-benzyl-3-[2-(tert-butoxy)-2-oxoethyl]-4-methylpyrrolidine-3-carboxylate

  • Molecular Weight : 423.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-1-benzyl-3-2-tert-butoxy-2-oxoethyl-4-methylpyrrolidine-3-carboxylate-308445.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85135
Availability details for
Quantity
  • Availability
  • Add to Cart