ADVH49B829C1 AChemBlock

tert-butyl (3aS,7aR)-rel-7,7-dimethyl-6-oxo-1,3,3a,4,5,7a-hexahydropyrrolo[3,4-c]pyridine-2-carboxylate

  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3as-7ar-rel-7-7-dimethyl-6-oxo-1-3-3a-4-5-7a-hexahydropyrrolo-3-4-c-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84763
Availability details for
Quantity
  • Availability
  • Add to Cart