ADVH49B829C3 AChemBlock

benzyl (3S)-3-amino-3-methylpiperidine-1-carboxylate

  • Molecular Weight : 248.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-3s-3-amino-3-methylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85139
Availability details for
Quantity
  • Availability
  • Add to Cart