ADVH49B829C6 AChemBlock

benzyl (2S,5S)-5-hydroxy-2-methyl-piperidine-1-carboxylate

  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2s-5s-5-hydroxy-2-methyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85142
Availability details for
Quantity
  • Availability
  • Add to Cart