ADVH49B829C7 AChemBlock

benzyl (2S,3R)-3-hydroxy-2-methyl-azetidine-1-carboxylate

  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2s-3r-3-hydroxy-2-methyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85143
Availability details for
Quantity
  • Availability
  • Add to Cart