ADVH49B829C8 AChemBlock

benzyl (2S)-2-ethynyl-4,4-difluoropyrrolidine-1-carboxylate

  • Molecular Weight : 265.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2s-2-ethynyl-4-4-difluoropyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85144
Availability details for
Quantity
  • Availability
  • Add to Cart