ADVH49B829CA AChemBlock

benzyl (2R,4R)-rel-4-amino-2-(trifluoromethyl)piperidine-1-carboxylate

  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2r-4r-rel-4-amino-2-trifluoromethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85146
Availability details for
Quantity
  • Availability
  • Add to Cart