ADVH49B829CE AChemBlock

benzyl (2R)-2-ethynyl-2-(trifluoromethyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 297.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2r-2-ethynyl-2-trifluoromethyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85150
Availability details for
Quantity
  • Availability
  • Add to Cart