ADVH49B829D1 AChemBlock

tert-butyl (3aS,6aS)-rel-3a-hydroxy-octahydropyrrolo[3,4-c]pyrrole-2-carboxylate

  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3as-6as-rel-3a-hydroxy-octahydropyrrolo-3-4-c-pyrrole-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84764
Availability details for
Quantity
  • Availability
  • Add to Cart