ADVH49B829D2 AChemBlock

9-tert-butyl 7-methyl (1R,5S)-3-oxa-9-azabicyclo[3.3.1]nonane-7,9-dicarboxylate

  • Molecular Weight : 285.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/9-tert-butyl-7-methyl-1r-5s-3-oxa-9-azabicyclo-3-3-1-nonane-7-9-dicarboxylate-308467.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85157
Availability details for
Quantity
  • Availability
  • Add to Cart