ADVH49B829DA AChemBlock

tert-butyl (3aR,7aS)-rel-3,3-difluoro-3a,4,5,6,7,7a-hexahydro-2H-pyrrolo[3,2-c]pyridine-1-carboxylate

  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3ar-7as-rel-3-3-difluoro-3a-4-5-6-7-7a-hexahydro-2h-pyrrolo-3-2-c-pyridine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84770
Availability details for
Quantity
  • Availability
  • Add to Cart