ADVH49B829DC AChemBlock

8-methyl-8-azabicyclo[3.2.1]octan-2-amine;dihydrochloride

  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-methyl-8-azabicyclo-3-2-1-octan-2-amine-dihydrochloride-308478.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85168
Availability details for
Quantity
  • Availability
  • Add to Cart