ADVH49B829DD AChemBlock

8-methyl-7-oxa-1-azaspiro[3.4]octane hydrochloride

  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-methyl-7-oxa-1-azaspiro-3-4-octane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85169
Availability details for
Quantity
  • Availability
  • Add to Cart