ADVH49B829DE AChemBlock

8-methoxy-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,2,4]triazolo[1,5-a]pyridine

  • Molecular Weight : 275.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-methoxy-6-tetramethyl-1-3-2-dioxaborolan-2-yl-1-2-4-triazolo-1-5-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85170
Availability details for
Quantity
  • Availability
  • Add to Cart