ADVH49B829E1 AChemBlock

8-ethoxycarbonyl-1,4-dioxaspiro[4.5]decane-8-carboxylic acid

  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-ethoxycarbonyl-1-4-dioxaspiro-4-5-decane-8-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85173
Availability details for
Quantity
  • Availability
  • Add to Cart