ADVH49B829E6 AChemBlock

tert-butyl (2S,6S)-4-hydroxy-2,6-dimethyl-piperidine-1-carboxylate

  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-6s-4-hydroxy-2-6-dimethyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84778
Availability details for
Quantity
  • Availability
  • Add to Cart