ADVH49B829E7 AChemBlock

tert-butyl (2S,6S)-2,6-dimethyl-3-oxo-piperazine-1-carboxylate

  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-6s-2-6-dimethyl-3-oxo-piperazine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84779
Availability details for
Quantity
  • Availability
  • Add to Cart