ADVH49B829E8 AChemBlock

8-[(benzyloxy)carbonyl]-2,8-diazaspiro[4.5]decane-3-carboxylic acid

  • Molecular Weight : 318.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-benzyloxy-carbonyl-2-8-diazaspiro-4-5-decane-3-carboxylic-acid-308490.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85180
Availability details for
Quantity
  • Availability
  • Add to Cart