ADVH49B829EA AChemBlock

8,8-difluoro-5-oxa-2-azaspiro[3.4]octane hydrochloride

  • Molecular Weight : 185.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-8-difluoro-5-oxa-2-azaspiro-3-4-octane-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85182
Availability details for
Quantity
  • Availability
  • Add to Cart