ADVH49B829F0 AChemBlock

7-oxabicyclo[2.2.1]heptan-2-amine hydrochloride

  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-oxabicyclo-2-2-1-heptan-2-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85190
Availability details for
Quantity
  • Availability
  • Add to Cart