ADVH49B829F7 AChemBlock

7-fluoro-1-methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

  • Molecular Weight : 276.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-fluoro-1-methyl-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85196
Availability details for
Quantity
  • Availability
  • Add to Cart