ADVH49B829F8 AChemBlock

7-ethylsulfonyl-4,7-diazaspiro[2.5]octane

  • Molecular Weight : 204.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-ethylsulfonyl-4-7-diazaspiro-2-5-octane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85197
Availability details for
Quantity
  • Availability
  • Add to Cart