ADVH49B829FE AChemBlock

tert-butyl (2S)-2-(bromomethyl)-2-methylpyrrolidine-1-carboxylate

  • Molecular Weight : 278.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-2-bromomethyl-2-methylpyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84788
Availability details for
Quantity
  • Availability
  • Add to Cart