ADVH49B82A01 AChemBlock

tert-butyl 5-(2-methoxy-2-oxo-ethyl)-2-azabicyclo[2.2.2]octane-2-carboxylate

  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-2-methoxy-2-oxo-ethyl-2-azabicyclo-2-2-2-octane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84611
Availability details for
Quantity
  • Availability
  • Add to Cart