ADVH49B82A04 AChemBlock

7-bromo-3,3-dimethyl-1H,2H,3H-pyrrolo[3,2-c]pyridin-2-one

  • Molecular Weight : 241.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-3-3-dimethyl-1h-2h-3h-pyrrolo-3-2-c-pyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85208
Availability details for
Quantity
  • Availability
  • Add to Cart