ADVH49B82A06 AChemBlock

7-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic aicd

  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1h-pyrrolo-3-2-c-pyridine-3-carboxylic-aicd.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72420
Availability details for
Quantity
  • Availability
  • Add to Cart