ADVH49B82A09 AChemBlock

7-{[(tert-butyldimethylsilyl)oxy]methyl}-3-iodoimidazo[1,2-a]pyridine

  • Molecular Weight : 388.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-tert-butyldimethylsilyl-oxy-methyl-3-iodoimidazo-1-2-a-pyridine-308522.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85212
Availability details for
Quantity
  • Availability
  • Add to Cart