ADVH49B82A0B AChemBlock

tert-butyl 3-(2-hydroxyethyl)-3-methyl-azetidine-1-carboxylate

  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-2-hydroxyethyl-3-methyl-azetidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84669
Availability details for
Quantity
  • Availability
  • Add to Cart