ADVH49B82A0D AChemBlock

7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolo[2,1-f][1,2,4]triazin-4-amine

  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrrolo-2-1-f-1-2-4-triazin-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85216
Availability details for
Quantity
  • Availability
  • Add to Cart