ADVH49B82A0E AChemBlock

tert-butyl (2R,3S)-3-hydroxy-2-vinyl-piperidine-1-carboxylate hydrochloride

  • Molecular Weight : 263.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-3s-3-hydroxy-2-vinyl-piperidine-1-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84795
Availability details for
Quantity
  • Availability
  • Add to Cart