ADVH49B82A10 AChemBlock

tert-butyl (2R,3S)-2-(3-bromopropyl)-3-[(tert-butyldimethylsilyl)oxy]piperidine-1-carboxylate

  • Molecular Weight : 436.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-3s-2-3-bromopropyl-3-tert-butyldimethylsilyl-oxy-piperidine-1-carboxylate-308106.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84796
Availability details for
Quantity
  • Availability
  • Add to Cart