ADVH49B82A11 AChemBlock

tert-butyl (2R)-4,4-difluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrolidine-1-carboxylate

  • Molecular Weight : 333.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-4-4-difluoro-2-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84797
Availability details for
Quantity
  • Availability
  • Add to Cart