ADVH49B82A14 AChemBlock

tert-butyl (2R)-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate

  • Molecular Weight : 311.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-2-tetramethyl-1-3-2-dioxaborolan-2-yl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84800
Availability details for
Quantity
  • Availability
  • Add to Cart