ADVH49B82A16 AChemBlock

tert-butyl 4-(bromomethyl)isoindoline-2-carboxylate

  • Molecular Weight : 312.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-bromomethyl-isoindoline-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84630
Availability details for
Quantity
  • Availability
  • Add to Cart