ADVH49B82A17 AChemBlock

6-methyl-6-azaspiro[3.4]octan-2-ol

  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-6-azaspiro-3-4-octan-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85229
Availability details for
Quantity
  • Availability
  • Add to Cart