ADVH49B82A1E AChemBlock

6-hydrazinyl-2-methyl-2-azaspiro[3.3]heptane trihydrochloride

  • Molecular Weight : 250.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-hydrazinyl-2-methyl-2-azaspiro-3-3-heptane-trihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85235
Availability details for
Quantity
  • Availability
  • Add to Cart