ADVH49B82A20 AChemBlock

6-fluoro-2,3,4,5-tetrahydro-1H-1-benzazepin-9-amine hydrochloride

  • Molecular Weight : 216.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-2-3-4-5-tetrahydro-1h-1-benzazepin-9-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85237
Availability details for
Quantity
  • Availability
  • Add to Cart