ADVH49B82A23 AChemBlock

tert-butyl 2-methyl-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine-5-carboxylate

  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-methyl-3h-4h-5h-6h-7h-imidazo-4-5-c-pyridine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84673
Availability details for
Quantity
  • Availability
  • Add to Cart