ADVH49B82A25 AChemBlock

6-chloropyrrolo[1,2-a]pyrazine-3-carboxylic acid

  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloropyrrolo-1-2-a-pyrazine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85241
Availability details for
Quantity
  • Availability
  • Add to Cart