ADVH49B82A2A AChemBlock

6-chloro-4-isopropyl-pyridazin-3-ol

  • Molecular Weight : 172.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-isopropyl-pyridazin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85246
Availability details for
Quantity
  • Availability
  • Add to Cart