ADVH49B82A2C AChemBlock

tert-butyl (1S,5R,6S)-6-hydroxy-3-azabicyclo[3.2.0]heptane-3-carboxylate

  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1s-5r-6s-6-hydroxy-3-azabicyclo-3-2-0-heptane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84807
Availability details for
Quantity
  • Availability
  • Add to Cart