ADVH49B82A2E AChemBlock

6-chloro-3-(4-chlorophenyl)-4-(4-pyridyl)pyridazine

  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3-4-chlorophenyl-4-4-pyridyl-pyridazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85250
Availability details for
Quantity
  • Availability
  • Add to Cart