ADVH49B82A31 AChemBlock

6-bromo-7-fluoro-5-methoxy-1H-indazole

  • Molecular Weight : 245.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-7-fluoro-5-methoxy-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85252
Availability details for
Quantity
  • Availability
  • Add to Cart