ADVH49B82A35 AChemBlock

6-bromo-5,7-dimethoxy-1H-indazole

  • Molecular Weight : 257.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-5-7-dimethoxy-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85255
Availability details for
Quantity
  • Availability
  • Add to Cart