ADVH49B82A42 AChemBlock

6,7-difluoroquinazoline-2-carboxylic acid

  • Molecular Weight : 210.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-7-difluoroquinazoline-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85265
Availability details for
Quantity
  • Availability
  • Add to Cart