ADVH49B82A43 AChemBlock

6,7,8,9-tetrahydro-5H-imidazo[1,2-a][1,4]diazepine;hydrochloride

  • Molecular Weight : 173.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-7-8-9-tetrahydro-5h-imidazo-1-2-a-1-4-diazepine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85266
Availability details for
Quantity
  • Availability
  • Add to Cart