ADVH49B82A44 AChemBlock

tert-butyl (1R,5S,6R)-rel-6-(aminomethyl)-3-azabicyclo[3.2.0]heptane-3-carboxylate

  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1r-5s-6r-rel-6-aminomethyl-3-azabicyclo-3-2-0-heptane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84816
Availability details for
Quantity
  • Availability
  • Add to Cart