ADVH49B82A45 AChemBlock

6-(trifluoromethyl)quinoxaline-2-carboxylic acid

  • Molecular Weight : 242.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-trifluoromethyl-quinoxaline-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85268
Availability details for
Quantity
  • Availability
  • Add to Cart